Product Details

SNP ID
rs1579255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:35819521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCATCTGTCTAGTTGTTTTACCC[A/G]TTATTGAAAGTAGGGTGTTGAAGGT
Phenotype
MIM: 603151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPT7 PubMed Links

Gene Details

Gene
SEPT7
Gene Name
septin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011553.3 Intron NP_001011553.2
NM_001242956.1 Intron NP_001229885.1
NM_001788.5 Intron NP_001779.3
XM_006715806.3 Intron XP_006715869.1
XM_011515656.2 Intron XP_011513958.1
XM_011515661.2 Intron XP_011513963.1
XM_017012855.1 Intron XP_016868344.1
XM_017012856.1 Intron XP_016868345.1
XM_017012857.1 Intron XP_016868346.1
XM_017012858.1 Intron XP_016868347.1
XM_017012859.1 Intron XP_016868348.1
XM_017012860.1 Intron XP_016868349.1
XM_017012861.1 Intron XP_016868350.1
XM_017012862.1 Intron XP_016868351.1
XM_017012863.1 Intron XP_016868352.1

View Full Product Details