Product Details

SNP ID
rs2271546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41221292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGAGGGGTCAGAGGACATGTGG[C/T]GCTCATAGGTTGTGGGAAGTCAGGA
Phenotype
MIM: 109135
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AXL PubMed Links

Gene Details

Gene
AXL
Gene Name
AXL receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278599.1 Intron NP_001265528.1
NM_001699.5 Intron NP_001690.2
NM_021913.4 Intron NP_068713.2

View Full Product Details