Product Details
- SNP ID
-
rs7902613
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:44961613 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAGCCCCCAACTCCCAGTGTCAA[C/T]AAGAATAGTAATGGGAGCCTCTTTT
- Phenotype
-
MIM: 610559
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RASSF4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77521840] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RASSF4
- Gene Name
- Ras association domain family member 4
- Gene
- TMEM72-AS1
- Gene Name
- TMEM72 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details