Product Details

SNP ID
rs3770415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:53674340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAACTTAGTTTATCCATAGATCT[A/G]TTAGCAGATGAGTAATATTTAACTT
Phenotype
MIM: 605760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB3 PubMed Links

Gene Details

Gene
ASB3
Gene Name
ankyrin repeat and SOCS box containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201965.1 Intron NP_001188894.1
NM_016115.4 Intron NP_057199.1
NM_145863.2 Intron NP_665862.1
Gene
GPR75-ASB3
Gene Name
GPR75-ASB3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164165.1 Intron NP_001157637.1

View Full Product Details