Product Details

SNP ID
rs34505194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:24952709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATTATACATATATTGACTTTT[C/T]AATTCACATAGAATCTGGAACTCAA
Phenotype
MIM: 162280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6841 PubMed Links

Gene Details

Gene
MIR6841
Gene Name
microRNA 6841
There are no transcripts associated with this gene.

Gene
NEFL
Gene Name
neurofilament, light polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006158.4 2087 UTR 3 NP_006149.2

View Full Product Details