Product Details

SNP ID
rs2236239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93936590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCATCACTGCCCCAGAAACTTTA[A/T]CAAAGGCTTCAATTCAAGTGGCTTC
Phenotype
MIM: 605759
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ASB2 PubMed Links

Gene Details

Gene
ASB2
Gene Name
ankyrin repeat and SOCS box containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202429.1 Intron NP_001189358.1
NM_016150.4 Intron NP_057234.2
XM_005267758.3 Intron XP_005267815.1
XM_011536834.2 Intron XP_011535136.1
XM_011536835.2 Intron XP_011535137.1
XM_017021369.1 Intron XP_016876858.1
Gene
FAM181A
Gene Name
family with sequence similarity 181 member A
There are no transcripts associated with this gene.

View Full Product Details