Product Details

SNP ID
rs8115115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59963807 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGAAATTCTGTGATTTCTAGGGT[C/T]CAAATTCTCCCTGAATGAAAGCTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH26 PubMed Links

Gene Details

Gene
CDH26
Gene Name
cadherin 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021810.4 Intron NP_068582.2
NM_177980.2 Intron NP_817089.1
XM_011528969.2 Intron XP_011527271.1
XM_011528970.2 Intron XP_011527272.1
XM_017027994.1 Intron XP_016883483.1
XM_017027995.1 Intron XP_016883484.1
XM_017027996.1 Intron XP_016883485.1
XM_017027997.1 Intron XP_016883486.1
XM_017027998.1 Intron XP_016883487.1
XM_017027999.1 Intron XP_016883488.1
XM_017028000.1 Intron XP_016883489.1
XM_017028001.1 Intron XP_016883490.1
XM_017028002.1 Intron XP_016883491.1

View Full Product Details