Product Details
- SNP ID
-
rs4148646
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:17393643 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGCTGTGCACTGATGACGGCCACAA[C/G]AGGCCAGTCCTGTCCCTGGGTGTCC
- Phenotype
-
MIM: 600509
MIM: 600937
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCC8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17846719] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCC8
- Gene Name
- ATP binding cassette subfamily C member 8
- Gene
- KCNJ11
- Gene Name
- potassium voltage-gated channel subfamily J member 11
There are no transcripts associated with this gene.
View Full Product Details