Product Details

SNP ID
rs590329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:144286171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTCCTCCCCGCCGGGGTGACAG[A/G]GTCGCCCTCCGTGCATCGCGCGGTG
Phenotype
MIM: 128240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UTRN PubMed Links

Gene Details

Gene
UTRN
Gene Name
utrophin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007124.2 Intron NP_009055.2
XM_005267127.4 Intron XP_005267184.1
XM_005267130.2 Intron XP_005267187.1
XM_005267133.2 Intron XP_005267190.1
XM_006715560.3 Intron XP_006715623.1
XM_011536101.2 Intron XP_011534403.1
XM_011536102.2 Intron XP_011534404.1
XM_011536106.2 Intron XP_011534408.1
XM_011536107.2 Intron XP_011534409.1
XM_011536109.2 Intron XP_011534411.1
XM_017011243.1 Intron XP_016866732.1
XM_017011244.1 Intron XP_016866733.1
XM_017011245.1 Intron XP_016866734.1

View Full Product Details