Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005490.2 | 1772 | UTR 3 | NP_005481.2 | ||
XM_005259467.1 | 1772 | UTR 3 | XP_005259524.1 | ||
XM_005259472.2 | 1772 | UTR 3 | XP_005259529.1 | ||
XM_006722602.1 | 1772 | UTR 3 | XP_006722665.1 | ||
XM_011527608.2 | 1772 | UTR 3 | XP_011525910.1 | ||
XM_011527609.2 | 1772 | UTR 3 | XP_011525911.1 | ||
XM_017026123.1 | 1772 | Intron | XP_016881612.1 | ||
XM_017026124.1 | 1772 | Intron | XP_016881613.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001288962.1 | 1772 | Intron | NP_001275891.1 | ||
NM_001288963.1 | 1772 | Intron | NP_001275892.1 | ||
NM_004240.3 | 1772 | Intron | NP_004231.1 | ||
XM_005259683.2 | 1772 | Intron | XP_005259740.1 | ||
XM_006722940.1 | 1772 | Intron | XP_006723003.1 |