Product Details

SNP ID
rs2608016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52273972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATGGATACACGTTATGACATAG[C/T]AATAAAGAAAAGCAAGGGAATGACC
Phenotype
MIM: 601928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT86 PubMed Links

Gene Details

Gene
KRT86
Gene Name
keratin 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320198.1 3466 Intron NP_001307127.1
XM_005268866.4 3466 UTR 5 XP_005268923.1
XM_017019296.1 3466 Intron XP_016874785.1

View Full Product Details