Product Details

SNP ID
rs13031561
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:210483606 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTCCCTGCTGGACATCAGGGCTCTT[A/T]TATGCTGCCTTGACTATATGGGTCC
Phenotype
MIM: 608307 MIM: 604155
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CPS1 PubMed Links

Gene Details

Gene
CPS1
Gene Name
carbamoyl-phosphate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122633.2 Intron NP_001116105.1
NM_001122634.3 Intron NP_001116106.1
NM_001875.4 Intron NP_001866.2
XM_011510640.1 Intron XP_011508942.1
XM_011510641.2 Intron XP_011508943.1
XM_011510642.2 Intron XP_011508944.1
XM_011510643.2 Intron XP_011508945.1
XM_011510644.2 Intron XP_011508946.1
Gene
LANCL1
Gene Name
LanC like 1
There are no transcripts associated with this gene.

View Full Product Details