Product Details
- SNP ID
-
rs12647895
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:39408562 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACAGTTGATCCTCTTAACAGCCTT[A/T]TGAACTAGTTTTATTATTCTTCCCA
- Phenotype
-
MIM: 611135
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLB
PubMed Links
Gene Details
- Gene
- KLB
- Gene Name
- klotho beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175737.3 |
|
Intron |
|
|
NP_783864.1 |
- Gene
- MIR5591
- Gene Name
- microRNA 5591
There are no transcripts associated with this gene.
View Full Product Details