Product Details
- SNP ID
-
rs13005816
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:31876525 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTATGACAGCAAGAACCATGTTTG[C/T]TTACTGCTGTATCTCCACCAACACC
- Phenotype
-
MIM: 611786
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MEMO1
PubMed Links
Gene Details
- Gene
- MEMO1
- Gene Name
- mediator of cell motility 1
There are no transcripts associated with this gene.
View Full Product Details