Product Details

SNP ID
rs11222364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130868695 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAACCCCCCGACCACACCCCATG[A/C]CCAGTATACTCTTAGAGCAGAGAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100507431 PubMed Links

Gene Details

Gene
LOC100507431
Gene Name
uncharacterized LOC100507431
There are no transcripts associated with this gene.

Gene
LOC103611081
Gene Name
uncharacterized LOC103611081
There are no transcripts associated with this gene.

Gene
SNX19
Gene Name
sorting nexin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301089.1 11483 Intron NP_001288018.1
NM_014758.2 11483 Intron NP_055573.2
XM_005271546.4 11483 UTR 3 XP_005271603.1
XM_011542819.2 11483 UTR 3 XP_011541121.1
XM_011542820.2 11483 UTR 3 XP_011541122.1
XM_011542821.2 11483 UTR 3 XP_011541123.1
XM_011542822.2 11483 Intron XP_011541124.1
XM_011542824.2 11483 UTR 3 XP_011541126.1
XM_011542825.2 11483 UTR 3 XP_011541127.1
XM_011542826.2 11483 UTR 3 XP_011541128.1
XM_011542827.2 11483 UTR 3 XP_011541129.1
XM_017017725.1 11483 UTR 3 XP_016873214.1
XM_017017726.1 11483 UTR 3 XP_016873215.1

View Full Product Details