Product Details

SNP ID
rs1749435
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:43166268 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAACCATCGCACTGTGTGATGGG[A/G]AATCAACTTTACTCAGCATTCACTT
Phenotype
MIM: 614259 MIM: 614443
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP57 PubMed Links

Gene Details

Gene
CFAP57
Gene Name
cilia and flagella associated protein 57
There are no transcripts associated with this gene.

Gene
EBNA1BP2
Gene Name
EBNA1 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159936.1 Intron NP_001153408.1
NM_006824.2 Intron NP_006815.2
Gene
FAM183A
Gene Name
family with sequence similarity 183 member A
There are no transcripts associated with this gene.

Gene
MIR6733
Gene Name
microRNA 6733
There are no transcripts associated with this gene.

View Full Product Details