Product Details
- SNP ID
-
rs291105
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
47
- Location
-
Chr.1:206906696 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GTTCCATCAAGATTTTTGCCCCTAA[T/G]AGTGGGAATGAGAGTTTCAAGTAGA
- Phenotype
-
MIM: 606015
MIM: 604136
- Polymorphism
- T/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FCMR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75522788] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FCMR
- Gene Name
- Fc fragment of IgM receptor
- Gene
- IL24
- Gene Name
- interleukin 24
There are no transcripts associated with this gene.
View Full Product Details