Product Details
- SNP ID
-
rs1079093
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
39
- Location
-
Chr.1:114093604 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAGAGCCAGAACCCGAAACCAGGT[C/T]TCCTAACACCCAACCAAGTGTTCTT
- Phenotype
-
MIM: 607718
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SYT6
PubMed Links
Gene Details
- Gene
- SYT6
- Gene Name
- synaptotagmin 6
View Full Product Details