Product Details

SNP ID
rs11083904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47878969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAATGAAGACACAGCGGGATGGA[A/G]GGAATTTAAGGAAAGTAAGGATGGT
Phenotype
MIM: 125263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01595 PubMed Links

Gene Details

Gene
LINC01595
Gene Name
long intergenic non-protein coding RNA 1595
There are no transcripts associated with this gene.

Gene
SULT2A1
Gene Name
sulfotransferase family 2A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003167.3 Intron NP_003158.2

View Full Product Details