Product Details

SNP ID
rs7297662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56001594 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGGGGTACTACTGTACTTGTTG[A/G]CTCTTCATGTTAGCTCTGCTAGGCA
Phenotype
MIM: 606239 MIM: 179514 MIM: 606887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IKZF4 PubMed Links

Gene Details

Gene
IKZF4
Gene Name
IKAROS family zinc finger 4
There are no transcripts associated with this gene.

Gene
LOC105369781
Gene Name
uncharacterized LOC105369781
There are no transcripts associated with this gene.

Gene
RAB5B
Gene Name
RAB5B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SUOX
Gene Name
sulfite oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000456.2 Intron NP_000447.2
NM_001032386.1 Intron NP_001027558.1
NM_001032387.1 Intron NP_001027559.1
XM_005269112.1 Intron XP_005269169.1
XM_017019905.1 Intron XP_016875394.1
XM_017019906.1 Intron XP_016875395.1
XM_017019907.1 Intron XP_016875396.1
XM_017019908.1 Intron XP_016875397.1

View Full Product Details