Product Details
- SNP ID
-
rs3734666
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:14120337 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCATTGATTATGCTTATACAACAAA[A/C]ATGGATAGAGTGTTCTGGAGACAAG
- Phenotype
-
MIM: 604534
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD83
PubMed Links
Gene Details
- Gene
- CD83
- Gene Name
- CD83 molecule
View Full Product Details