Product Details
- SNP ID
-
rs3808978
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:90133019 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTACAAAAGTTTCAAGAAATAAGT[C/T]ATTAGGCATACCTATCAACAATTAT
- Phenotype
-
MIM: 611636
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NAALAD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112174672] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NAALAD2
- Gene Name
- N-acetylated alpha-linked acidic dipeptidase 2
View Full Product Details