Product Details

SNP ID
rs568700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:128858038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCTCAGTGTTCTTTTTCATAAT[A/G]AGGATGCCAATATGGGTTGTGAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLT1D1 PubMed Links

Gene Details

Gene
GLT1D1
Gene Name
glycosyltransferase 1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144669.2 Intron NP_653270.1
XM_005253553.4 Intron XP_005253610.2
XM_011537956.2 Intron XP_011536258.2
XM_011537957.2 Intron XP_011536259.1
XM_017018855.1 Intron XP_016874344.1
XM_017018856.1 Intron XP_016874345.1
XM_017018857.1 Intron XP_016874346.1
XM_017018858.1 Intron XP_016874347.1
XM_017018859.1 Intron XP_016874348.1
XM_017018860.1 Intron XP_016874349.1
XM_017018861.1 Intron XP_016874350.1
Gene
LOC100128276
Gene Name
uncharacterized LOC100128276
There are no transcripts associated with this gene.

Gene
LOC101929984
Gene Name
uncharacterized LOC101929984
There are no transcripts associated with this gene.

View Full Product Details