Product Details

SNP ID
rs10141798
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:94582313 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCTCATAGATCAGGAAACCGAAGC[A/T]TGAAGAATACAGGCGGCAGATGCCT
Phenotype
MIM: 601841
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SERPINA5 PubMed Links

Gene Details

Gene
SERPINA5
Gene Name
serpin family A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000624.5 Intron NP_000615.3

View Full Product Details