Product Details

SNP ID
rs11910271
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:32772234 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGGGAGGAGGAGGAGGGGAGAGGCC[A/G]GAGAGAGAAGCGGCTTTCGGGAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C21orf62-AS1 PubMed Links

Gene Details

Gene
C21orf62-AS1
Gene Name
C21orf62 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PAXBP1
Gene Name
PAX3 and PAX7 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013329.3 Intron NP_037461.2
NM_016631.3 Intron NP_057715.2
XM_006724066.1 Intron XP_006724129.1
XM_011529804.2 Intron XP_011528106.1

View Full Product Details