Product Details

SNP ID
rs7268
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:140332965 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCCAGAAAGGACCATGACAGCATA[A/C]CTTCATCGTTTTGGTGAGGTGGGTG
Phenotype
MIM: 126150
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HBEGF PubMed Links

Gene Details

Gene
HBEGF
Gene Name
heparin binding EGF like growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001945.2 2236 UTR 3 NP_001936.1

View Full Product Details