Product Details

SNP ID
rs1055131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119834883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAACTTGAGTCCTGAAGAAAAGAC[C/T]GTTGTTCTTTTAAGAAATGGGTCTT
Phenotype
MIM: 613203 MIM: 604912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSCC1 PubMed Links

Gene Details

Gene
DSCC1
Gene Name
DNA replication and sister chromatid cohesion 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024094.2 1327 UTR 3 NP_076999.2
XM_005251065.3 1327 UTR 3 XP_005251122.1
Gene
TAF2
Gene Name
TATA-box binding protein associated factor 2
There are no transcripts associated with this gene.

View Full Product Details