Product Details

SNP ID
rs1582957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:21750299 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTTGCATCTTTTTATGTTTTCCA[A/C]GCTAGTGTGCAATCCGCTTGAAAGC
Phenotype
MIM: 600562
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDH12 PubMed Links

Gene Details

Gene
CDH12
Gene Name
cadherin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317227.1 4537 Intron NP_001304156.1
NM_001317228.1 4537 Intron NP_001304157.1
NM_004061.4 4537 Intron NP_004052.2
XM_011513927.2 4537 UTR 3 XP_011512229.1
XM_017008920.1 4537 UTR 3 XP_016864409.1
XM_017008921.1 4537 UTR 3 XP_016864410.1

View Full Product Details