Product Details

SNP ID
rs2005488
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:36499225 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGCCATTAGAGCTGTCAGTGACCT[G/C]GCTGCAGCAGCACCACAGTTGCTGT
Phenotype
MIM: 604500
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
MYO19 PubMed Links

Gene Details

Gene
MYO19
Gene Name
myosin XIX
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033580.2 1979 Intron NP_001028752.1
NM_001163735.1 1979 Intron NP_001157207.1
NM_025109.5 1979 Intron NP_079385.2
XM_011525286.1 1979 Intron XP_011523588.1
XM_011525287.1 1979 Intron XP_011523589.1
XM_011525290.2 1979 Intron XP_011523592.1
XM_017025157.1 1979 Intron XP_016880646.1
XM_017025158.1 1979 Intron XP_016880647.1
XM_017025159.1 1979 Intron XP_016880648.1
XM_017025160.1 1979 Intron XP_016880649.1
XM_017025161.1 1979 Intron XP_016880650.1
Gene
ZNHIT3
Gene Name
zinc finger HIT-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281432.1 1979 UTR 3 NP_001268361.1
NM_001281433.1 1979 Intron NP_001268362.1
NM_001281434.1 1979 Intron NP_001268363.1
NM_004773.3 1979 Intron NP_004764.1

View Full Product Details