Product Details

SNP ID
rs1050156
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87094700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCAGCAACGCGGCCGAGTCGG[C/T]GGACGCCGAGCCCAGGGCAGCGGGC
Phenotype
MIM: 138130
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM35A PubMed Links

Gene Details

Gene
FAM35A
Gene Name
family with sequence similarity 35 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019054.2 223 Intron NP_061927.2
XM_005269920.3 223 Intron XP_005269977.1
XM_006717903.3 223 Intron XP_006717966.1
XM_006717905.3 223 Intron XP_006717968.1
XM_011539876.2 223 Intron XP_011538178.1
XM_011539877.2 223 Intron XP_011538179.1
XM_011539878.2 223 Intron XP_011538180.1
XM_011539879.2 223 Intron XP_011538181.1
XM_011539881.2 223 Intron XP_011538183.1
XM_011539882.2 223 Intron XP_011538184.1
XM_011539883.2 223 Intron XP_011538185.1
XM_017016349.1 223 Intron XP_016871838.1
XM_017016350.1 223 UTR 5 XP_016871839.1
XM_017016351.1 223 Intron XP_016871840.1
XM_017016352.1 223 Intron XP_016871841.1
XM_017016353.1 223 Intron XP_016871842.1
Gene
GLUD1
Gene Name
glutamate dehydrogenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318900.1 223 Intron NP_001305829.1
NM_001318901.1 223 Intron NP_001305830.1
NM_001318902.1 223 Intron NP_001305831.1
NM_001318904.1 223 UTR 5 NP_001305833.1
NM_001318905.1 223 UTR 5 NP_001305834.1
NM_001318906.1 223 UTR 5 NP_001305835.1
NM_005271.4 223 Missense Mutation ACC,GCC T24A NP_005262.1

View Full Product Details