Product Details

SNP ID
rs999797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30832311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGCAGGCCGGGCTGGACCGCGT[A/G]AGGTCCTAGGAGACGGGATTCCGGG
Phenotype
MIM: 609534 MIM: 614853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATAD5 PubMed Links

Gene Details

Gene
ATAD5
Gene Name
ATPase family, AAA domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024857.4 363 UTR 5 NP_079133.3
XM_011525269.2 363 UTR 5 XP_011523571.1
XM_011525270.2 363 UTR 5 XP_011523572.1
XM_011525273.2 363 UTR 5 XP_011523575.1
XM_011525274.2 363 Intron XP_011523576.1
XM_017025111.1 363 Intron XP_016880600.1
Gene
CRLF3
Gene Name
cytokine receptor like factor 3
There are no transcripts associated with this gene.

View Full Product Details