Product Details
- SNP ID
-
rs1633550
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:113798483 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTTGGGCCAGCAGTAGCCCTATTGA[C/G]TGTTTTTGACAATTTTTAGGTATTA
- Phenotype
-
MIM: 610748
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC390251
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114628994] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC390251
- Gene Name
- SH3 domain containing GRB2 like 1 pseudogene
There are no transcripts associated with this gene.
- Gene
- USP28
- Gene Name
- ubiquitin specific peptidase 28
View Full Product Details