Product Details

SNP ID
rs198756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61622528 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTAACAACTGCTCCCTCAGTTC[C/T]GCACAGAGAAGGCTGTTCCCAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPLP0P2 PubMed Links

Gene Details

Gene
RPLP0P2
Gene Name
ribosomal protein lateral stalk subunit P0 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details