Product Details

SNP ID
rs1018783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109098052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAATCCGCGCGCCGCAGGCCCTCC[A/T]GGCTCGGTGCGCTGTCCAATCAGAG
Phenotype
MIM: 610602 MIM: 191525
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALKBH2 PubMed Links

Gene Details

Gene
ALKBH2
Gene Name
alkB homolog 2, alpha-ketoglutarate dependent dioxygenase
There are no transcripts associated with this gene.

Gene
UNG
Gene Name
uracil DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003362.3 Intron NP_003353.1
NM_080911.2 Intron NP_550433.1

View Full Product Details