Product Details

SNP ID
rs10555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:107168886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACCTTTTCTATGGATTTCCTT[C/G]ATCTCTCAGTCACACTGCAAACTTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM155A PubMed Links

Gene Details

Gene
FAM155A
Gene Name
family with sequence similarity 155 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080396.2 3504 UTR 3 NP_001073865.1
XM_011521109.2 3504 Intron XP_011519411.1

View Full Product Details