Product Details
- SNP ID
-
rs1009910
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:74945395 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCGCGCGGGCCCAGAAAAGAGATGA[A/G]AGCATAAACCCCTAGGGGGCGCAGT
- Phenotype
-
MIM: 601121
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PGF
PubMed Links
Gene Details
- Gene
- PGF
- Gene Name
- placental growth factor
View Full Product Details