Product Details

SNP ID
rs1181574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:43765096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGCAGACCGGCTGCAGTAGATA[C/T]GGGGGGAGGAGAAACCTGCTTTAAA
Phenotype
MIM: 109750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BLVRA PubMed Links

Gene Details

Gene
BLVRA
Gene Name
biliverdin reductase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000712.3 Intron NP_000703.2
NM_001253823.1 Intron NP_001240752.1
XM_011515474.2 Intron XP_011513776.1
XM_017012520.1 Intron XP_016868009.1

View Full Product Details