Product Details

SNP ID
rs1052145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103630116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGATGCTCCTCTCGGAGGAGC[G/T]CCCTCCGCAGGAGGGTCTTTCCAGG
Phenotype
MIM: 300237
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TCEAL1 PubMed Links

Gene Details

Gene
TCEAL1
Gene Name
transcription elongation factor A like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006639.1 362 Missense Mutation CGC,CTC R67L NP_001006640.1
NM_001006640.1 362 Missense Mutation CGC,CTC R67L NP_001006641.1
NM_004780.2 362 Missense Mutation CGC,CTC R67L NP_004771.2

View Full Product Details