Product Details

SNP ID
rs1529856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11288522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGAGGAAGCTAGAACTGTGAGT[A/G]TCTGGGGGAAGAGCATTGCAGCCAG
Phenotype
MIM: 606260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTMR9 PubMed Links

Gene Details

Gene
MTMR9
Gene Name
myotubularin related protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015458.3 Intron NP_056273.2
XM_011543830.2 Intron XP_011542132.1
XM_011543831.2 Intron XP_011542133.1
XM_017013753.1 Intron XP_016869242.1

View Full Product Details