Product Details
- SNP ID
-
rs10103397
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:71199364 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTACAGGTACTCCAGTTCCAAGGC[A/G]TGGTGCAGGGCCATGAGGTCCGAGT
- Phenotype
-
MIM: 601653
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EYA1
PubMed Links
Gene Details
- Gene
- EYA1
- Gene Name
- EYA transcriptional coactivator and phosphatase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000503.5 |
2395 |
Silent Mutation |
CAC,CAT |
H585H |
NP_000494.2 |
NM_001288574.1 |
2395 |
Silent Mutation |
CAC,CAT |
H579H |
NP_001275503.1 |
NM_001288575.1 |
2395 |
Silent Mutation |
CAC,CAT |
H463H |
NP_001275504.1 |
NM_172058.3 |
2395 |
Silent Mutation |
CAC,CAT |
H585H |
NP_742055.1 |
NM_172059.3 |
2395 |
Silent Mutation |
CAC,CAT |
H550H |
NP_742056.1 |
NM_172060.3 |
2395 |
Silent Mutation |
CAC,CAT |
H552H |
NP_742057.1 |
XM_011517483.2 |
2395 |
Silent Mutation |
CAC,CAT |
H584H |
XP_011515785.1 |
XM_011517484.2 |
2395 |
Silent Mutation |
CAC,CAT |
H609H |
XP_011515786.2 |
XM_017013201.1 |
2395 |
Silent Mutation |
CAC,CAT |
H614H |
XP_016868690.1 |
XM_017013202.1 |
2395 |
Silent Mutation |
CAC,CAT |
H614H |
XP_016868691.1 |
XM_017013203.1 |
2395 |
Silent Mutation |
CAC,CAT |
H613H |
XP_016868692.1 |
XM_017013204.1 |
2395 |
Silent Mutation |
CAC,CAT |
H608H |
XP_016868693.1 |
XM_017013205.1 |
2395 |
Intron |
|
|
XP_016868694.1 |
XM_017013206.1 |
2395 |
Silent Mutation |
CAC,CAT |
H585H |
XP_016868695.1 |
XM_017013207.1 |
2395 |
Silent Mutation |
CAC,CAT |
H584H |
XP_016868696.1 |
XM_017013208.1 |
2395 |
Silent Mutation |
CAC,CAT |
H584H |
XP_016868697.1 |
XM_017013209.1 |
2395 |
Silent Mutation |
CAC,CAT |
H579H |
XP_016868698.1 |
XM_017013210.1 |
2395 |
Silent Mutation |
CAC,CAT |
H578H |
XP_016868699.1 |
XM_017013211.1 |
2395 |
Silent Mutation |
CAC,CAT |
H564H |
XP_016868700.1 |
XM_017013212.1 |
2395 |
Silent Mutation |
CAC,CAT |
H552H |
XP_016868701.1 |
XM_017013213.1 |
2395 |
Silent Mutation |
CAC,CAT |
H468H |
XP_016868702.1 |
View Full Product Details