Product Details

SNP ID
rs10103644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:71199475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAGCACCCAGCGCCAGCCCTGC[C/T]AGCTTTTTTGGAAATCCACTCCCAT
Phenotype
MIM: 601653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EYA1 PubMed Links

Gene Details

Gene
EYA1
Gene Name
EYA transcriptional coactivator and phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000503.5 Intron NP_000494.2
NM_001288574.1 Intron NP_001275503.1
NM_001288575.1 Intron NP_001275504.1
NM_172058.3 Intron NP_742055.1
NM_172059.3 Intron NP_742056.1
NM_172060.3 Intron NP_742057.1
XM_011517483.2 Intron XP_011515785.1
XM_011517484.2 Intron XP_011515786.2
XM_017013201.1 Intron XP_016868690.1
XM_017013202.1 Intron XP_016868691.1
XM_017013203.1 Intron XP_016868692.1
XM_017013204.1 Intron XP_016868693.1
XM_017013205.1 Intron XP_016868694.1
XM_017013206.1 Intron XP_016868695.1
XM_017013207.1 Intron XP_016868696.1
XM_017013208.1 Intron XP_016868697.1
XM_017013209.1 Intron XP_016868698.1
XM_017013210.1 Intron XP_016868699.1
XM_017013211.1 Intron XP_016868700.1
XM_017013212.1 Intron XP_016868701.1
XM_017013213.1 Intron XP_016868702.1

View Full Product Details