Product Details

SNP ID
rs2391391
Assay Type
Validated
NCBI dbSNP Submissions
71
Location
Chr.1:94986370 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAGAAGCAAGGGAGTATAGAAGGA[A/G]TGGGTCTTTGCTTACCCTATATTGA
Phenotype
MIM: 612866
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG14 PubMed Links

Gene Details

Gene
ALG14
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305242.1 Intron NP_001292171.1
NM_144988.3 Intron NP_659425.1
XM_005270582.3 Intron XP_005270639.1
XM_011540897.2 Intron XP_011539199.1

View Full Product Details