Product Details

SNP ID
rs2919878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88071922 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCTAGCTTCTACATAGCATGGGG[T/C]CAAACTGGGAATTCGCAATGCTGGG
Phenotype
MIM: 606846
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SMYD1 PubMed Links

Gene Details

Gene
SMYD1
Gene Name
SET and MYND domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198274.3 Intron NP_938015.1
XM_005264156.2 Intron XP_005264213.1

View Full Product Details