Product Details

SNP ID
rs1130902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:182890893 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTTTCCCTCAACTCTGCAGCAAA[C/T]ACTTCAATGCTACTGTCATTTGAGG
Phenotype
MIM: 607638
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCTD PubMed Links

Gene Details

Gene
DCTD
Gene Name
dCMP deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012732.1 1310 UTR 3 NP_001012750.1
NM_001921.2 1310 UTR 3 NP_001912.2
XM_005262778.3 1310 UTR 3 XP_005262835.1
XM_005262779.3 1310 UTR 3 XP_005262836.1
XM_005262780.3 1310 UTR 3 XP_005262837.1
XM_005262781.4 1310 UTR 3 XP_005262838.1
XM_005262782.3 1310 UTR 3 XP_005262839.1
XM_006714115.3 1310 UTR 3 XP_006714178.1
XM_006714116.3 1310 UTR 3 XP_006714179.1
XM_011531674.2 1310 UTR 3 XP_011529976.1
XM_011531675.2 1310 UTR 3 XP_011529977.1
XM_011531676.2 1310 UTR 3 XP_011529978.1
XM_017007816.1 1310 UTR 3 XP_016863305.1
XM_017007817.1 1310 UTR 3 XP_016863306.1
XM_017007818.1 1310 UTR 3 XP_016863307.1
XM_017007819.1 1310 UTR 3 XP_016863308.1
XM_017007820.1 1310 UTR 3 XP_016863309.1
XM_017007821.1 1310 UTR 3 XP_016863310.1
XM_017007822.1 1310 UTR 3 XP_016863311.1
XM_017007823.1 1310 UTR 3 XP_016863312.1
XM_017007824.1 1310 UTR 3 XP_016863313.1

View Full Product Details