Product Details

SNP ID
rs1960206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:182892218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCCCACTGCTGCTGAACGCCAA[C/T]ACCATGAAGCGCTGGAAAACTTTAC
Phenotype
MIM: 607638
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCTD PubMed Links

Gene Details

Gene
DCTD
Gene Name
dCMP deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012732.1 Intron NP_001012750.1
NM_001921.2 Intron NP_001912.2
XM_005262778.3 Intron XP_005262835.1
XM_005262779.3 Intron XP_005262836.1
XM_005262780.3 Intron XP_005262837.1
XM_005262781.4 Intron XP_005262838.1
XM_005262782.3 Intron XP_005262839.1
XM_006714115.3 Intron XP_006714178.1
XM_006714116.3 Intron XP_006714179.1
XM_011531674.2 Intron XP_011529976.1
XM_011531675.2 Intron XP_011529977.1
XM_011531676.2 Intron XP_011529978.1
XM_017007816.1 Intron XP_016863305.1
XM_017007817.1 Intron XP_016863306.1
XM_017007818.1 Intron XP_016863307.1
XM_017007819.1 Intron XP_016863308.1
XM_017007820.1 Intron XP_016863309.1
XM_017007821.1 Intron XP_016863310.1
XM_017007822.1 Intron XP_016863311.1
XM_017007823.1 Intron XP_016863312.1
XM_017007824.1 Intron XP_016863313.1

View Full Product Details