Product Details
- SNP ID
-
rs1203843
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:98849730 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTGGGAAGAACTAACTCCTACCCC[A/G]GGACCAGGAATCTGAAACCCTGGCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM130
PubMed Links
Gene Details
- Gene
- TMEM130
- Gene Name
- transmembrane protein 130
View Full Product Details