Product Details

SNP ID
rs1288630
Assay Type
Functionally Tested
NCBI dbSNP Submissions
30
Location
Chr.1:53461200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAGCTCGGCACTGAGGCCTCA[C/G]AGAGGCAGAGTGCGGGCTGACGGTG
Phenotype
MIM: 614805
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DMRTB1 PubMed Links

Gene Details

Gene
DMRTB1
Gene Name
DMRT like family B with proline rich C-terminal 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033067.2 Intron NP_149056.1

View Full Product Details