Product Details

SNP ID
rs536551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:128859268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGCAAGGTGGGAGAACGGAGGG[A/G]GAGGCGTCTTAGGCAGAGCGTCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLT1D1 PubMed Links

Gene Details

Gene
GLT1D1
Gene Name
glycosyltransferase 1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144669.2 Intron NP_653270.1
XM_005253553.4 Intron XP_005253610.2
XM_011537956.2 Intron XP_011536258.2
XM_011537957.2 Intron XP_011536259.1
XM_017018855.1 Intron XP_016874344.1
XM_017018856.1 Intron XP_016874345.1
XM_017018857.1 Intron XP_016874346.1
XM_017018858.1 Intron XP_016874347.1
XM_017018859.1 Intron XP_016874348.1
XM_017018860.1 Intron XP_016874349.1
XM_017018861.1 Intron XP_016874350.1
Gene
LOC100128276
Gene Name
uncharacterized LOC100128276
There are no transcripts associated with this gene.

View Full Product Details