Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004914.3 | 38 | UTR 5 | NP_004905.2 | ||
XM_005261859.4 | 38 | UTR 5 | XP_005261916.1 | ||
XM_006724381.3 | 38 | UTR 5 | XP_006724444.1 | ||
XM_006724382.3 | 38 | UTR 5 | XP_006724445.1 | ||
XM_011530544.2 | 38 | UTR 5 | XP_011528846.1 | ||
XM_011530545.2 | 38 | UTR 5 | XP_011528847.1 | ||
XM_011530546.2 | 38 | UTR 5 | XP_011528848.1 | ||
XM_011530547.2 | 38 | UTR 5 | XP_011528849.1 | ||
XM_011530550.2 | 38 | UTR 5 | XP_011528852.1 | ||
XM_011530552.2 | 38 | Intron | XP_011528854.1 | ||
XM_017029104.1 | 38 | UTR 5 | XP_016884593.1 | ||
XM_017029105.1 | 38 | UTR 5 | XP_016884594.1 | ||
XM_017029106.1 | 38 | Intron | XP_016884595.1 | ||
XM_017029107.1 | 38 | Intron | XP_016884596.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014433.2 | 38 | Intron | NP_055248.1 | ||
XM_011530149.2 | 38 | Intron | XP_011528451.1 | ||
XM_011530150.1 | 38 | Intron | XP_011528452.1 | ||
XM_011530151.2 | 38 | Intron | XP_011528453.1 | ||
XM_011530152.2 | 38 | Intron | XP_011528454.1 | ||
XM_011530154.2 | 38 | Intron | XP_011528456.1 | ||
XM_011530155.2 | 38 | Intron | XP_011528457.1 | ||
XM_017028774.1 | 38 | Intron | XP_016884263.1 |