Product Details

SNP ID
rs1804885
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:45012955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATGACACATTCCTGCGAGCAGC[C/T]TGGGGAGAGGAAACAGACTACACTC
Phenotype
MIM: 613521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HECTD3 PubMed Links

Gene Details

Gene
HECTD3
Gene Name
HECT domain E3 ubiquitin protein ligase 3
There are no transcripts associated with this gene.

Gene
UROD
Gene Name
uroporphyrinogen decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000374.4 157 Silent Mutation GCC,GCT A23A NP_000365.3
XM_005271169.1 157 UTR 5 XP_005271226.1
XM_005271170.1 157 UTR 5 XP_005271227.1
Gene
ZSWIM5
Gene Name
zinc finger SWIM-type containing 5
There are no transcripts associated with this gene.

View Full Product Details